accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 605644-46-0

IUPAC Name: 9,9-dimethyl-2-nitro-9H-fluorene
Synonyms: 9,9-dimethyl-2-nitrofluorene
Molecular Weight (g/mol): 239.27
Molecular Formula: C15H13NO2
InChi Key: SPNVINZCDHRVAI-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results