accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 606-12-2

OHOOH

CAS RN 606-12-2

IUPAC Name: 4-(2-hydroxybenzoyl)phenol
Synonyms: 4-(2-hydroxybenzoyl)phenol
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: HUYKZYIAFUBPAQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1O
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results