accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 606-28-0

IUPAC Name: methyl 2-benzoylbenzoate
Synonyms: methyl 2-benzoylbenzoate
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: NQSMEZJWJJVYOI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)