accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 606130-90-9

IUPAC Name: 13-azido-2,5,8,11-tetraoxatridecane
Synonyms: 13-azido-2,5,8,11-tetraoxatridecane
Molecular Weight (g/mol): 233.27
Molecular Formula: C9H19N3O4
InChi Key: FFOZZVDSANUDAE-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCN=[N+]=[N-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results