accessibility menu, dialog, popup

UserName

Viewing profile as user:
FCeFFF

CAS RN 60627-09-0

IUPAC Name: tetrafluorocerium
Synonyms: ceric fluoride
Molecular Weight (g/mol): 216.11
Molecular Formula: CeF4
InChi Key: SWURHZJFFJEBEE-UHFFFAOYSA-J
SMILES: F[Ce](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results