accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 6064-38-6

IUPAC Name: pentadecan-7-one
Synonyms: pentadecan-7-one
Molecular Weight (g/mol): 226.4
Molecular Formula: C15H30O
InChi Key: VUXXXYMMZBAUSE-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results