accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 6064-90-0

IUPAC Name: methyl henicosanoate
Synonyms: methyl henicosanoate
Molecular Weight (g/mol): 340.59
Molecular Formula: C22H44O2
InChi Key: AJRICDSAJQHDSD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCC(=O)OC
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)