accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60662-54-6

IUPAC Name: cyclohexane-1,3,5-triol dihydrate
Synonyms: cyclohexane-1,3,5-triol dihydrate
Molecular Weight (g/mol): 168.19
Molecular Formula: C6H16O5
InChi Key: MAPLMYMZLRIJSP-UHFFFAOYSA-N
SMILES: O.O.OC1CC(O)CC(O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results