accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60698-32-0

IUPAC Name: 5-phenoxyfuran-2-carboxylic acid
Synonyms: 5-phenoxyfuran-2-carboxylic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: QQKXTFLRAZKJJO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=CC=C2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results