accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 607-71-6

IUPAC Name: 4-methyl-2H-chromen-2-one
Synonyms: 4-methyl-2H-chromen-2-one
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: PSGQCCSGKGJLRL-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results