accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 607-97-6

IUPAC Name: ethyl 2-ethyl-3-oxobutanoate
Synonyms: ethyl 2-ethyl-3-oxobutanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: OKANYBNORCUPKZ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CC)C(C)=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)