accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrBrBr

CAS RN 607-99-8

IUPAC Name: 1,3,5-tribromo-2-methoxybenzene
Synonyms: 2,4,6-tribromoanisole
Molecular Weight (g/mol): 344.83
Molecular Formula: C7H5Br3O
InChi Key: YXTRCOAFNXQTKL-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(Br)C=C1Br
Boiling Point 
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)