accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60702-69-4

IUPAC Name: 2-chloro-4-fluorobenzonitrile
Synonyms: 2-chloro-4-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: PGKPNNMOFHNZJX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)