accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OHOHCH3H3CCH3HOCH3CH3CH3OHCH3CH3H3C

CAS RN 60705-62-6

IUPAC Name: 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Synonyms: 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular Weight (g/mol): 648.93
Molecular Formula: C44H56O4
InChi Key: NVKLTRSBZLYZHK-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (3)
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (1)
  • (7)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)