accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3CSClOO

CAS RN 60706-63-0

IUPAC Name: 2,3,5,6-tetramethylbenzene-1-sulfonyl chloride
Synonyms: 2,3,5,6-tetramethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 232.72
Molecular Formula: C10H13ClO2S
InChi Key: ZCXRROBIIMQMHR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(=C1C)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)