accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHBr

CAS RN 60710-39-6

IUPAC Name: 3-bromo-4-methylphenol
Synonyms: 3-bromo-4-methylphenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: GMZKNRDHSHYMHG-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results