accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOONCH3CH3SOOO(Z)(E/Z)

CAS RN 60711-74-2

IUPAC Name: 3-octadecyl-2-{3-[(2Z)-3-octadecyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium 4-methylbenzene-1-sulfonate
Synonyms: 3-octadecyl-2-{3-[(2Z)-3-octadecyl-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium tosylate
Molecular Weight (g/mol): 953.47
Molecular Formula: C60H92N2O5S
InChi Key: AGONTKXFFSJLQO-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.CCCCCCCCCCCCCCCCCCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CCCCCCCCCCCCCCCCCC)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results