accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOH

CAS RN 6072-49-7

IUPAC Name: 2-(thiophen-2-yl)benzoic acid
Synonyms: 2-(thiophen-2-yl)benzoic acid
Molecular Weight (g/mol): 204.24
Molecular Formula: C11H8O2S
InChi Key: GDSOQCSYONDNAJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results