accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 607366-20-1

CH3CH3OOH3CH3CHNOHONHOO

CAS RN 607366-20-1

IUPAC Name: 3-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 532.64
Molecular Formula: C31H36N2O6
InChi Key: HLIFXCXTXPXGNH-UHFFFAOYNA-N
SMILES: CC(C)CC(NCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)