accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60758-86-3

IUPAC Name: 3-ethoxy-4-methoxybenzonitrile
Synonyms: 3-ethoxy-4-methoxybenzonitrile
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: XTIINWPNAMHVDG-UHFFFAOYSA-N
SMILES: CCOC1=C(OC)C=CC(=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results