accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 608-23-1

IUPAC Name: 1-chloro-2,3-dimethylbenzene
Synonyms: 1-chloro-2,3-dimethylbenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: NVLHGZIXTRYOKT-UHFFFAOYSA-N
SMILES: CC1=C(C)C(Cl)=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)