accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHHO

CAS RN 608-25-3

IUPAC Name: 2-methylbenzene-1,3-diol
Synonyms: 2-methylresorcinol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: ZTMADXFOCUXMJE-UHFFFAOYSA-N
SMILES: CC1=C(O)C=CC=C1O
Molecular Weight (g/mol) 
  • (8)
  • (4)
Percent Purity 
  • (5)
  • (7)
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
  • (2)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (8)
  • (4)
  • (5)
  • (7)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)