CAS RN 60811-17-8
IUPAC Name:
5-bromo-2-chloroaniline
Synonyms:
5-bromo-2-chloroaniline
Molecular Weight (g/mol):
206.47
Molecular Formula:
C6H5BrClN
InChi Key:
UGOLEPGQWYPIBR-UHFFFAOYSA-N
SMILES:
NC1=CC(Br)=CC=C1Cl