accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6082-66-2

IUPAC Name: 3,4,6-trichloropyridazine
Synonyms: 3,4,6-trichloropyridazine
Molecular Weight (g/mol): 183.42
Molecular Formula: C4HCl3N2
InChi Key: LJDQXQOPXOLCHL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)N=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results