accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OH2NHCl

CAS RN 6084-58-8

IUPAC Name: O-(2-methylpropyl)hydroxylamine hydrochloride
Synonyms: O-(2-methylpropyl)hydroxylamine hydrochloride
Molecular Weight (g/mol): 125.6
Molecular Formula: C4H12ClNO
InChi Key: WOXGREIMSJFDPG-UHFFFAOYSA-N
SMILES: Cl.CC(C)CON
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)