accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3H3CCH3N

CAS RN 608534-41-4

IUPAC Name: 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]-9H-carbazole
Synonyms: 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole
Molecular Weight (g/mol): 321.23
Molecular Formula: C20H24BNO2
InChi Key: NTUZDLWSQJRXAM-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCN2C3=C(C=CC=C3)C3=C2C=CC=C3)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results