accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)OHO(S)NH2H3COHCl

CAS RN 60856-51-1

IUPAC Name: (2S,3R)-2-amino-3-(benzyloxy)butanoic acid hydrochloride
Synonyms: O-benzyl-L-threonine hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: XECHCUXLJZQHOJ-SCYNACPDSA-N
SMILES: Cl.C[C@@H](OCC1=CC=CC=C1)[C@H](N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)