accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6088-88-6

IUPAC Name: [(2E)-but-2-en-1-yl](chloro)magnesium
Synonyms: (2E)-but-2-en-1-yl(chloro)magnesium
Molecular Weight (g/mol): 114.86
Molecular Formula: C4H7ClMg
InChi Key: RTJGBPYEBKZQGE-CZEFNJPISA-M
SMILES: C\C=C\C[Mg]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results