accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 609-08-5

IUPAC Name: 1,3-diethyl 2-methylpropanedioate
Synonyms: 1,3-diethyl 2-methylpropanedioate
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: UPQZOUHVTJNGFK-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)C(=O)OCC
Boiling Point 
  • (5)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)