accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrCH3CH3

CAS RN 609-12-1

IUPAC Name: ethyl 2-bromo-3-methylbutanoate
Synonyms: ethyl 2-bromo-3-methylbutanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: WNFUWONOILPKNX-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)C(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)