accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClCl

CAS RN 609-19-8

IUPAC Name: 3,4,5-trichlorophenol
Synonyms: 3,4,5-trichlorophenol
Molecular Weight (g/mol): 197.44
Molecular Formula: C6H3Cl3O
InChi Key: GBNHEBQXJVDXSW-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results