accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3SHOOO

CAS RN 609-54-1

IUPAC Name: 2,5-dimethylbenzene-1-sulfonic acid
Synonyms: 2,5-dimethylbenzenesulfonic acid
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: IRLYGRLEBKCYPY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)S(O)(=O)=O
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)