accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 609-71-2

IUPAC Name: 2-oxo-1,2-dihydropyridine-3-carboxylic acid
Synonyms: 2-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: UEYQJQVBUVAELZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CNC1=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)