accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3C

CAS RN 609-72-3

IUPAC Name: N,N,2-trimethylaniline
Synonyms: N,N-dimethyl-O-toluidine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: JDEJGVSZUIJWBM-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)