accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6092-80-4

IUPAC Name: hydrogen O-phenylhydroxylamine chloride
Synonyms: hydrogen O-aminophenol chloride
Molecular Weight (g/mol): 145.59
Molecular Formula: C6H8ClNO
InChi Key: DBTXKJJSFWZJNS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOC1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results