accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6094-02-6

IUPAC Name: 2-methylhex-1-ene
Synonyms: 2-methyl-1-hexene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: IRUDSQHLKGNCGF-UHFFFAOYSA-N
SMILES: CCCCC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results