accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONN

CAS RN 6094-60-6

IUPAC Name: 1-benzyl-4-hydroxypiperidine-4-carbonitrile
Synonyms: 1-benzyl-4-hydroxypiperidine-4-carbonitrile
Molecular Weight (g/mol): 216.28
Molecular Formula: C13H16N2O
InChi Key: XQRLXUYZKZXSBN-UHFFFAOYSA-N
SMILES: OC1(CCN(CC2=CC=CC=C2)CC1)C#N
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)