accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOH3COCH3

CAS RN 6095-42-7

IUPAC Name: trihexyl phosphite
Synonyms: trihexyl phosphite
Molecular Weight (g/mol): 334.48
Molecular Formula: C18H39O3P
InChi Key: DSKYTPNZLCVELA-UHFFFAOYSA-N
SMILES: CCCCCCOP(OCCCCCC)OCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results