accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrNOO

CAS RN 60956-26-5

IUPAC Name: 4-bromo-1-methyl-2-nitrobenzene
Synonyms: 4-bromo-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: KZNXALJXBRSMFL-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1[N+]([O-])=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)