accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrOH3C

CAS RN 60965-26-6

IUPAC Name: 2-bromo-1-(2,4-dimethoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 259.1
Molecular Formula: C10H11BrO3
InChi Key: PKVBZABQCCQHLD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(=O)CBr)C(OC)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)