CAS RN 6097-08-1
IUPAC Name:
2-(hex-5-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
2-(hex-5-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol):
227.26
Molecular Formula:
C14H13NO2
InChi Key:
WALVSJIWVWDFCU-UHFFFAOYSA-N
SMILES:
O=C1N(CCCCC#C)C(=O)C2=CC=CC=C12
1
–
1
of
1
results
1