accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOSO

CAS RN 6099-21-4

IUPAC Name: [2-(benzenesulfinyl)ethanesulfinyl]benzene
Synonyms: [2-(benzenesulfinyl)ethanesulfinyl]benzene
Molecular Weight (g/mol): 278.38
Molecular Formula: C14H14O2S2
InChi Key: CSZMCUCIIIAHQD-UHFFFAOYNA-N
SMILES: O=S(CCS(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results