accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCS

CAS RN 6099-88-3

IUPAC Name: 1-chloro-2-isothiocyanatoethane
Synonyms: 1-chloro-2-isothiocyanatoethane
Molecular Weight (g/mol): 121.58
Molecular Formula: C3H4ClNS
InChi Key: ZUWFBQUHBOUPFK-UHFFFAOYSA-N
SMILES: ClCCN=C=S
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)