accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OCH3NONH2NH2NHNOHOHSOHOO

CAS RN 61-47-2

IUPAC Name: 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one 3-(2-aminoethyl)-1H-indol-5-ol sulfuric acid hydrate
Synonyms: 1-methylhydantoin-2-imide serotonin sulfuric acid hydrate
Molecular Weight (g/mol): 405.43
Molecular Formula: C14H23N5O7S
InChi Key: BKCXVJIGPVULPX-UHFFFAOYSA-N
SMILES: O.OS(O)(=O)=O.CN1CC(=O)N=C1N.NCCC1=CNC2=CC=C(O)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)