accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClO

CAS RN 610-14-0

IUPAC Name: 2-nitrobenzoyl chloride
Synonyms: 2-nitrobenzoyl chloride
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: BWWHTIHDQBHTHP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results