accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610-15-1

IUPAC Name: 2-nitrobenzamide
Synonyms: 2-nitrobenzamide
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: KLGQWSOYKYFBTR-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)