accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3CNOO

CAS RN 610-22-0

IUPAC Name: 1,2-dimethyl 4-nitrobenzene-1,2-dicarboxylate
Synonyms: 1,2-dimethyl 4-nitrophthalate
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: XWBDWELWBUWSNI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1C(=O)OC)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results