accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONOONOO

CAS RN 610-30-0

IUPAC Name: 2,4-dinitrobenzoic acid
Synonyms: 2,4-dinitrobenzoic acid
Molecular Weight (g/mol): 212.12
Molecular Formula: C7H4N2O6
InChi Key: ZIIGSRYPZWDGBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results