accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610-34-4

IUPAC Name: ethyl 2-nitrobenzoate
Synonyms: ethyl 2-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: CPNMAYYYYSWTIV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results