accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOHO

CAS RN 610-35-5

IUPAC Name: 4-hydroxybenzene-1,2-dicarboxylic acid
Synonyms: 4-hydroxyphthalic acid
Molecular Weight (g/mol): 182.13
Molecular Formula: C8H6O5
InChi Key: MWRVRCAFWBBXTL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)